Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035052.1 | NP_001030224.1 | 4 - 5 | 411 | 79 | |
GenBank mRNA | BC102175.1 | - | 4 - 5 | 411 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |