Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040471.1 | NP_001035561.1 | 17 - 18 | 1620 | 87 | |
GenBank mRNA | BC103416.1 | - | 17 - 18 | 1620 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |