Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040471.1 | NP_001035561.1 | 1 - 2 | 200 | 56 | |
GenBank mRNA | BC103416.1 | - | 1 - 2 | 200 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |