Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077881.1 | NP_001071349.1 | 3 - 4 | 466 | 128 | |
GenBank mRNA | BC103143.1 | - | 3 - 4 | 466 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |