Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173897.1 | NP_776322.1 | 1 - 2 | 94 | 106 | |
GenBank mRNA | BC142534.1 | - | 1 - 2 | 109 | 106 | |
U58870.1 | - | 1 - 2 | 94 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |