Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173911.2 | NP_776336.1 | 4 - 5 | 416 | 153 | |
GenBank mRNA | BC102629.1 | - | 4 - 5 | 439 | 153 | |
X15556.1 | - | 416 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |