Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173967.3 | NP_776392.1 | 1 - 2 | 82 | 65 | |
GenBank mRNA | AB009591.1 | - | 1 - 2 | 82 | 65 | |
BC103035.1 | - | 1 - 2 | 99 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |