Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173980.2 | NP_776405.1 | 1 - 2 | 27 | 100 | |
GenBank mRNA | AF541976.1 | - | 1 - 2 | 27 | 100 | |
AJ011680.1 | - | 1 - 2 | 27 | 100 | ||
BC102211.1 | - | 1 - 2 | 49 | 100 | ||
BT029909.1 | - | 1 - 2 | 37 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |