Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174034.2 | NP_776459.1 | 1 - 2 | 85 | 106 | |
GenBank mRNA | AF036096.1 | - | 1 - 2 | 85 | 106 | |
BC142380.1 | - | 1 - 2 | 91 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |