Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314006.1 | NP_001300935.1 | 3 - 4 | 705 | 67 | |
NM_174055.3 | NP_776480.1 | 3 - 4 | 563 | 67 | ||
GenBank mRNA | BC103225.1 | - | 3 - 4 | 697 | 67 | |
EH374181.1 | - | 3 - 4 | 477 | 67 | ||
M35608.1 | - | 3 - 4 | 365 | 67 | ||
X13221.1 | - | 1192 | 67 | |||
X14032.1 | - | 365 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |