Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174067.2 | NP_776492.1 | 1 - 2 | 112 | 61 | |
GenBank mRNA | AF350329.1 | - | 1 - 2 | 112 | 61 | |
BC148027.1 | - | 2 - 3 | 156 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |