Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174070.2 | NP_776495.1 | 1 - 2 | 574 | 68 | |
GenBank mRNA | AF003927.2 | - | 1 - 2 | 574 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |