Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174093.1 | NP_776518.1 | 5 - 6 | 532 | 69 | |
GenBank mRNA | EU276067.1 | - | 4 - 5 | 487 | 69 | |
M35589.1 | - | 5 - 6 | 503 | 69 | ||
M37211.1 | - | 5 - 6 | 532 | 69 | ||
X12498.1 | - | 503 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |