Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174144.2 | NP_776569.1 | 1 - 2 | 492 | 67 | |
GenBank mRNA | AF259078.1 | - | 1 - 2 | 492 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |