Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174189.3 | NP_776614.2 | 1 - 2 | 198 | 89 | |
GenBank mRNA | BC110213.1 | - | 1 - 2 | 199 | 89 | |
Y17259.1 | - | 198 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |