Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316955.1 | NP_001303884.1 | 2 - 3 | 1164 | 59 | |
NM_001316956.1 | NP_001303885.1 | 2 - 3 | 1164 | 59 | ||
NM_001316992.1 | NP_001303921.1 | 2 - 3 | 1164 | 59 | ||
NM_001316993.1 | NP_001303922.1 | 2 - 3 | 1164 | 59 | ||
NM_174216.2 | NP_776641.1 | 2 - 3 | 1164 | 59 | ||
GenBank mRNA | AB450824.1 | - | 2 - 3 | 117 | 59 | |
AB455252.1 | - | 2 - 3 | 117 | 59 | ||
GU362062.1 | - | 2 - 3 | 117 | 59 | ||
GU362063.1 | - | 2 - 3 | 117 | 59 | ||
M31836.1 | - | 1 - 2 | 39 | 59 | ||
M32976.1 | - | 2 - 3 | 662 | 59 | ||
M33750.1 | - | 1 - 2 | 39 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |