Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174290.2 | NP_776715.1 | 1 - 2 | 208 | 74 | |
GenBank mRNA | AF029793.2 | - | 1 - 2 | 208 | 74 | |
BC102745.1 | - | 1 - 2 | 255 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |