Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077402.1 | NP_001070870.1 | 2 - 3 | 70 | 90 | |
NM_174317.2 | NP_776742.1 | 2 - 3 | 77 | 90 | ||
GenBank mRNA | AF132036.1 | - | 2 - 3 | 77 | 90 | |
BC112756.1 | - | 2 - 3 | 70 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |