Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174323.2 | NP_776748.1 | 6 - 7 | 1459 | 146 | |
GenBank mRNA | D90335.1 | - | 6 - 7 | 1459 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |