Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174326.2 | NP_776751.1 | 2 - 3 | 330 | 95 | |
GenBank mRNA | M11116.1 | - | 2 - 3 | 330 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |