Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174358.2 | NP_776783.1 | 6 - 7 | 933 | 152 | |
GenBank mRNA | BC133546.1 | - | 6 - 7 | 901 | 152 | |
M20818.1 | - | 6 - 7 | 933 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |