Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LALBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174378.2 | NP_776803.1 | 3 - 4 | 397 | 115 | |
GenBank mRNA | BC102173.1 | - | 3 - 4 | 397 | 115 | |
BT025469.1 | - | 3 - 4 | 381 | 115 | ||
FJ232912.1 | - | 3 - 4 | 370 | 115 | ||
J05147.1 | - | 3 - 4 | 397 | 115 | ||
M18780.1 | - | 3 - 4 | 378 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |