Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174400.2 | NP_776825.1 | 7 - 8 | 828 | 64 | |
GenBank mRNA | AB008852.1 | - | 7 - 8 | 828 | 64 | |
BC146237.1 | - | 7 - 8 | 850 | 64 | ||
BC151569.1 | - | 7 - 8 | 850 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |