Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174543.2 | NP_776968.1 | 5 - 6 | 611 | 134 | |
GenBank mRNA | BC123424.1 | - | 5 - 6 | 740 | 134 | |
X61456.1 | - | 1 | 611 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |