Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1SW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174567.1 | NP_776992.1 | 3 - 4 | 691 | 75 | |
GenBank mRNA | U92557.1 | - | 3 - 4 | 691 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Bos taurus Probe spans exons |