Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174634.1 | NP_777059.1 | 2 - 3 | 300 | 70 | |
GenBank mRNA | X53556.1 | - | 300 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |