Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174701.2 | NP_777126.1 | 6 - 7 | 545 | 84 | |
GenBank mRNA | AF202997.1 | - | 6 - 7 | 545 | 84 | |
AF238859.1 | - | 6 - 7 | 545 | 84 | ||
BC133473.1 | - | 6 - 7 | 551 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |