Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113255.1 | NP_001106726.1 | 3 - 4 | 257 | 81 | |
NM_174705.1 | NP_777130.1 | 1 - 2 | 224 | 81 | ||
GenBank mRNA | M33617.1 | - | 1 - 2 | 224 | 81 | |
M33618.1 | - | 3 - 4 | 257 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |