Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174728.2 | NP_777153.1 | 4 - 5 | 452 | 149 | |
GenBank mRNA | AF169160.1 | - | 4 - 5 | 452 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |