Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174800.2 | NP_777225.1 | 4 - 5 | 635 | 106 | |
GenBank mRNA | D84513.1 | - | 4 - 5 | 635 | 106 | |
D84514.1 | - | 168 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Inventoried Probe spans exons |