Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175800.2 | NP_786994.1 | 2 - 3 | 188 | 122 | |
GenBank mRNA | BC102292.1 | - | 2 - 3 | 189 | 122 | |
X63215.1 | - | 188 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |