Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175806.2 | NP_787000.1 | 4 - 5 | 580 | 65 | |
GenBank mRNA | BC104511.1 | - | 4 - 5 | 548 | 65 | |
U10039.1 | - | 4 - 5 | 580 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |