Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175806.2 | NP_787000.1 | 9 - 10 | 1222 | 79 | |
GenBank mRNA | BC104511.1 | - | 9 - 10 | 1190 | 79 | |
U10039.1 | - | 9 - 10 | 1222 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |