Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176609.3 | NP_788782.2 | 2 - 3 | 224 | 69 | |
GenBank mRNA | BC122610.1 | - | 2 - 3 | 176 | 69 | |
DV824401.1 | - | 2 - 3 | 223 | 69 | ||
EH148795.1 | - | 2 - 3 | 256 | 69 | ||
X56685.1 | - | 173 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |