Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176620.2 | NP_788793.1 | 3 - 4 | 359 | 74 | |
GenBank mRNA | AF020511.1 | - | 3 - 4 | 359 | 74 | |
BC123608.1 | - | 3 - 4 | 368 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |