Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177485.2 | NP_803451.1 | 4 - 5 | 464 | 75 | |
GenBank mRNA | BC114815.1 | - | 4 - 5 | 75 | ||
M59799.1 | - | 4 - 5 | 464 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |