Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181004.1 | NP_851347.1 | 1 - 2 | 50 | 109 | |
GenBank mRNA | AF347679.1 | - | 1 - 2 | 50 | 109 | |
X93304.1 | - | 50 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |