Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181006.2 | NP_851349.1 | 4 - 5 | 1969 | 58 | |
GenBank mRNA | U88551.1 | - | 4 - 5 | 1969 | 58 | |
X55027.1 | - | 1977 | 58 | |||
X55489.1 | - | 1965 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |