Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181022.2 | NP_851365.1 | 2 - 3 | 282 | 102 | |
GenBank mRNA | K03253.1 | - | 2 - 3 | 209 | 102 | |
K03254.1 | - | 2 - 3 | 238 | 102 | ||
X02440.1 | - | 282 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |