Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181031.2 | NP_851374.1 | 1 - 2 | 224 | 70 | |
GenBank mRNA | BC147979.1 | - | 1 - 2 | 313 | 70 | |
BT021727.1 | - | 1 - 2 | 103 | 70 | ||
X52554.1 | - | 224 | 70 | |||
X72858.1 | - | 262 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |