Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205776.1 | NP_991345.1 | 11 - 12 | 1060 | 83 | |
GenBank mRNA | AF241776.1 | - | 11 - 12 | 1060 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |