Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205776.1 | NP_991345.1 | 3 - 4 | 250 | 108 | |
GenBank mRNA | AF241776.1 | - | 3 - 4 | 250 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |