Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205802.2 | NP_991371.2 | 2 - 3 | 447 | 103 | |
GenBank mRNA | AF276819.1 | - | 2 - 3 | 398 | 103 | |
BC149605.1 | - | 2 - 3 | 445 | 103 | ||
EH143146.1 | - | 2 - 3 | 445 | 103 | ||
EV638847.1 | - | 2 - 3 | 156 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |