Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206970.1 | NP_996853.1 | 1 - 2 | 83 | 73 | |
GenBank mRNA | AY525122.1 | - | 1 - 2 | 83 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Inventoried Probe spans exons |