Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012519.1 | NP_001012537.1 | 8 - 9 | 848 | 70 | |
GenBank mRNA | BC110178.1 | - | 8 - 9 | 905 | 70 | |
BT021058.1 | - | 8 - 9 | 848 | 70 | ||
M31463.1 | - | 7 - 8 | 764 | 70 | ||
S54973.1 | - | 8 - 9 | 795 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |