Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013582.1 | NP_001013600.1 | 4 - 5 | 276 | 77 | |
GenBank mRNA | BC114796.1 | - | 4 - 5 | 316 | 77 | |
BT020951.1 | - | 4 - 5 | 276 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |