Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014388.1 | NP_001014410.1 | 1 - 2 | 127 | 82 | |
GenBank mRNA | AB098973.1 | - | 1 - 2 | 117 | 82 | |
AB099016.1 | - | 1 - 2 | 116 | 82 | ||
AB099031.1 | - | 1 - 2 | 131 | 82 | ||
BC104562.1 | - | 1 - 2 | 136 | 82 | ||
BT021036.1 | - | 1 - 2 | 127 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |