Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014388.1 | NP_001014410.1 | 2 - 3 | 203 | 67 | |
GenBank mRNA | AB098973.1 | - | 2 - 3 | 193 | 67 | |
AB099016.1 | - | 2 - 3 | 192 | 67 | ||
AB099031.1 | - | 2 - 3 | 207 | 67 | ||
BC104562.1 | - | 2 - 3 | 212 | 67 | ||
BT021036.1 | - | 2 - 3 | 203 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |