Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014855.1 | NP_001014855.1 | 2 - 3 | 76 | 100 | |
GenBank mRNA | BT020983.1 | - | 2 - 3 | 76 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |