Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014857.1 | NP_001014857.1 | 2 - 3 | 390 | 62 | |
NM_001105609.2 | NP_001099079.1 | 2 - 3 | 599 | 62 | ||
GenBank mRNA | BC153270.1 | - | 2 - 3 | 600 | 62 | |
BT020910.1 | - | 2 - 3 | 390 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |